Learn More
Sino Biological Recombinant Human IL-1 beta Protein, HPLC-verified
Supplier:  Sino Biological 10139HNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1β (NP_000567.1) (Ala 117-Ser 269) was expressed with a N-terminal Met.
Catalog No. 50-237-1294