Learn More
Sino Biological Anti-IL1R1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10126MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1R1 (rh IL1R1; Catalog#10126-H08H; NP_000868.1; Met 1-Thr 332). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8598