Learn More
Sino Biological Recombinant Human IL1R1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10126H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL1R1 (NP_000868.1) (Met 1-Thr 332) was expressed with a fused polyhistidine tag at C-terminus.
Catalog No. 50-255-5458