Learn More
Sino Biological Recombinant Human IL1RAPL1 Protein (hFc Tag)
Supplier:  Sino Biological 10177H02H100
Encompass_Preferred
A DNA sequence encoding the human IL1R8 (NP_055086.1) extracellular domain (Met 1-Leu 354) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6863