Learn More
BIOHIPPO Human IL1RA Microsample
SDP
Catalog No. 5000313848
Encompass
Human IL1RA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1RA in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL1RA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL1RA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL1RA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13848 Supplier BIOHIPPO Supplier No. BHE15212152
Human IL1RA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1RA in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL1RA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL1RA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL1RA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color