Learn More
BIOHIPPO Human IL1RAP Microsample
SDP
Catalog No. 5000315005
Encompass
Human IL1RAP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1RAP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL1RAP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL1RAP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL1RAP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15005 Supplier BIOHIPPO Supplier No. BHE15211789
Human IL1RAP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1RAP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL1RAP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL1RAP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL1RAP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color