Learn More
Sino Biological Human IL1RAPL2 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 40263T6220
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1R9 (NP_059112.1) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4165