Learn More
BIOHIPPO Human IL1RL1 Microsample
SDP
Catalog No. 5000314501
Encompass
Human IL1RL1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1RL1 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 22 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL1RL1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL1RL1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL1RL1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14501 Supplier BIOHIPPO Supplier No. BHE15211425
Human IL1RL1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1RL1 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 22 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL1RL1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL1RL1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL1RL1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color