Learn More
Sino Biological Anti-ST2/IL-1 RL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10105RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL1RL1 extracellular domain (rhIL1RL1; Catalog#10105-H08H; Met1-Phe328; NP_003847.2). IL1RL1 specific IgG was purified by human IL1RL1 affinity chromatography.
Catalog No. 50-240-8780