Learn More
Sino Biological Recombinant Human IL1RL2 Protein (His Tag) 1
Supplier:  Sino Biological 14398H08H1
Encompass_Preferred
A DNA sequence encoding the human IL1RL2 (NP_003845.2) (Met1-Tyr337) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7105