Learn More
Sino Biological Human IL1RL2 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 14398H02H1
Encompass_Preferred
A DNA sequence encoding the human IL1RL2 (NP_003845.2) (Met1-Tyr337) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1398