Learn More
Sino Biological Human IL2 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11848MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-2 (rh IL-2; Catalog#11848-HNAE; P60568-1; Ala21-Thr153). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-237-3167