Learn More
Sino Biological Anti-IL-2 Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 11848T16B20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL2/IL-2/Interleukin-2. IL2/IL-2/Interleukin-2 specific IgG was purified by Human IL2/IL-2/Interleukin-2 affinity chromatography and then biotinylated.
Catalog No. 50-241-2396