Learn More
Sino Biological Recombinant Human IL-2 Protein, HPLC-verified
Supplier:  Sino Biological 11848HNAE250
Encompass_Preferred
A DNA sequence encoding the mature form of human IL2 (P60568-1) (Ala21-Thr153) was expressed and purified with an initial Met.
Catalog No. 50-228-2650