Learn More
Sino Biological Human IL2 Protein 500ug (Sino Biological)
Supplier:  Sino Biological GMP11848HNAE500
Encompass_Preferred
A DNA sequence encoding the mature form of human IL2 (P60568-1) (Ala 21-Thr 153) was expressed and purified with an initial Met.
Catalog No. 50-237-1031