Learn More
Sino Biological Recombinant Anti-IL-20 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13060R04420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human interleukin-20 (rh interleukin-20; Catalog#13060-HNAE; Q9NYY1; Leu25-Glu176).
Catalog No. 50-255-4324