Learn More
Sino Biological Recombinant Human IL-20 Protein
Supplier:  Sino Biological 13060HNAE1
Encompass_Preferred
A DNA sequence encoding the mature form of human IL20 (Q9NYY1) (Leu25-Glu176) was expressed, with a N-terminal Met.
Catalog No. 50-255-4327