Learn More
Sino Biological Human IL21 PAb 20ul (Sino Biological)
Supplier:  Sino Biological 10584T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-21 (rh IL-21; Catalog#10584-HNAE; Q9HBE4-1; Gln30-Ser162). IL-21 specific IgG was purified by Human IL-21 affinity chromatography.
Catalog No. 50-237-1054