Learn More
Bon Opus Biosciences Recombinant Human IL-21R (C-His) - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP19150UG
Encompass_Preferred
Recombinant Human Interleukin-21 Receptor is produced by our Mammalian expression system and the target gene encoding Cys20-Pro236 is expressed with a 6His tag at the C-terminus. Accession#Q9HBE5. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH7.4
Catalog No. NC2733653