Learn More
Sino Biological Anti-IL-21 Receptor/IL-21R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11483RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-21R / Il21R (rh IL21R; Catalog#11483-H08H; Q9HBE5-1; Met1-Pro236). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8274