Learn More
Sino Biological Recombinant Human IL-21 Receptor/IL-21R Protein (hFc Tag)
Supplier:  Sino Biological 11483H02H100
Encompass_Preferred
A DNA sequence encoding the human IL21R (NP_068570.1) (Met1-Pro236) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4709