Learn More
Sino Biological Human IL22 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370470
Encompass_Preferred
Each vial contains 8.8 ng of recombinant Human IL22. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 130 pg/mL is recommended.
Catalog No. 50-237-0470 Supplier Sino Biological Supplier No. SEK1305915
Each vial contains 8.8 ng of recombinant Human IL22. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 130 pg/mL is recommended.