Learn More
Sino Biological Human IL22 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 13059MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL22 / Interleukin 22 (rh IL22 / Interleukin 22; Catalog#13059-HNAE; NP_065386.1; Ala34-Ile179). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1326