Learn More
Sino Biological Recombinant Human IL-22 Protein
Supplier:  Sino Biological 13059HNAE1
Encompass_Preferred
A DNA sequence encoding human IL22(NP_065386.1) (Ala34-Ile179) was expressed with a N-terminal Met.
Catalog No. 50-255-4328