Learn More
Sino Biological Recombinant Human IL-22BP Protein (His Tag) 1 mg
Supplier:  Sino Biological 10781H08H1
Encompass_Preferred
A DNA sequence encoding the human IL22RA2 (NP_443194.1) (Met1-Pro263) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7103