Learn More
Sino Biological Recombinant Anti-IL-22BP Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11025R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-22BP/IL-22RA2 (rh IL-22BP/IL-22RA2; Catalog#11025-H08H; NP_851826.1; Met1-Pro231).
Catalog No. 50-241-1777