Learn More
Sino Biological Recombinant Anti-IL-22BP Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11025R02820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL22RA2 / IL22BP (rh IL22BP; Catalog#11025-H08H; NP_851826.1; Met 1-Pro 231).
Catalog No. 50-240-9461