Learn More
BIOHIPPO Human IL22Ra2 Microsample
SDP
Catalog No. 5000315132
Encompass
Human IL22Ra2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL22Ra2 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 135 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL22Ra2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL22Ra2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL22Ra2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15132 Supplier BIOHIPPO Supplier No. BHE15211727
Human IL22Ra2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL22Ra2 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 135 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL22Ra2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL22Ra2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL22Ra2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color