Learn More
Sino Biological Recombinant Human IL-23 Protein (His Tag)
Supplier:  Sino Biological CT048H08H500
Encompass_Preferred
A DNA sequence encoding the human IL23 complex composed of IL12B (NP_002178.2)(Met1-Ser328) and the mature form of human IL23A (NP_057668.1) (Arg20-Pro189) linked by a polypeptide linker was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7035