Learn More
ACROBiosystems Human IL-23A&IL-12B Heterodimer Protein (ILB-H5219) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (IL-23A) & Arg 20 - Pro 189 (IL-12B) (Accession # Q9NPF7-1 (IL23A) & P29460-1 (IL12B)).
Catalog No. 502253334
Encompass_Preferred
Interleukin-23 subunit alpha (IL-23 alpha) can associates with IL12B to form the IL-23 interleukin, a heterodimeric cytokine which functions in innate and adaptive immunity. IL-23 may constitute with IL-17 an acute response to infection in peripheral tissues. IL-23 binds to a heterodimeric receptor complex composed of IL12RB1 and IL23R, activates the Jak-Stat signaling cascade, stimulates memory rather than naive T-cells and promotes production of proinflammatory cytokines. IL-23 induces autoimmune inflammation and thus may be responsible for autoimmune inflammatory diseases and may be important for tumorigenesis.
This item is not returnable. View return policy
Catalog No. 50-225-3334 Supplier ACROBiosystems Supplier No. ILBH521920UG
Interleukin-23 subunit alpha (IL-23 alpha) can associates with IL12B to form the IL-23 interleukin, a heterodimeric cytokine which functions in innate and adaptive immunity. IL-23 may constitute with IL-17 an acute response to infection in peripheral tissues. IL-23 binds to a heterodimeric receptor complex composed of IL12RB1 and IL23R, activates the Jak-Stat signaling cascade, stimulates memory rather than naive T-cells and promotes production of proinflammatory cytokines. IL-23 induces autoimmune inflammation and thus may be responsible for autoimmune inflammatory diseases and may be important for tumorigenesis.