Learn More
Sino Biological Recombinant Human IL-25/IL-17E Protein (hFc Tag) 500 µg
Supplier:  Sino Biological 10096H01H500
Encompass_Preferred
A DNA sequence encoding the mature form of human IL25 (Tyr 33-Gly177) (NP_073626.1) was fused with the Fc region of human IgG1 at the N-terminus via a polypeptide linker.
Catalog No. 50-224-7005