Learn More
Sino Biological Anti-IL-27 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10076RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-27 (rh IL-27; Catalog#10076-H08S; NP_005746.2 & NP_663634.2; Met1-Lys229 & Phe29-Pro243). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8646