Learn More
Sino Biological Anti-CD25/IL-2R alpha Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10165MM1420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD25 / IL2RA (rh CD25 / IL2RA; Catalog#10165-H08H; NP_000408.1; Met1-Cys213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1497