Learn More
Sino Biological Anti-CD25/IL-2R alpha Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 10165MM17TC25
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-2340