Learn More
Sino Biological Recombinant Human CD25/IL-2R alpha Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10165H02H1
Encompass_Preferred
A DNA sequence encoding the amino acid residues (Met 1-Cys 213) of the human IL2 receptor α chain (NP_000408.1) precursor was expressed with C-terminal fused human IgG1 Fc region.
Catalog No. 50-255-5449