Learn More
Sino Biological Recombinant Human CD25/IL-2R alpha Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10165H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domainof human IL2RA (NP_000408.1) (Met1-Cys213) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-5448