Learn More
Sino Biological Recombinant Human IL-2R beta/CD122 Protein (ECD, mFc Tag)
Supplier:  Sino Biological 10696H05H100
Encompass_Preferred
A DNA sequence encoding the human IL2RB (NP_000869.1) (Met1-Asp239) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-255-5207