Learn More
BIOHIPPO Human IL2Rg Microsample
SDP
Catalog No. 5000315013
Encompass
Human IL2Rg Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL2Rg in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL2Rg Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL2Rg Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL2Rg biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15013 Supplier BIOHIPPO Supplier No. BHE15211464
Human IL2Rg Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL2Rg in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL2Rg Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL2Rg Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL2Rg biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color