Learn More
Sino Biological Anti-IL-2R gamma/CD132 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10555T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD132 / IL2RG (rh CD132 / IL2RG; Catalog#10555-H08B; P31785); Met1-Asn254). CD132 / IL2RG specific IgG was purified by Human CD132 / IL2RG affinity chromatography.
Catalog No. 50-241-1503