Learn More
Sino Biological Anti-IL-2R gamma/CD132 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10555RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD132 / IL2RG (rh IL2RG; Catalog#10555-H08B; P31785); Met1-Asn254). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8221