Learn More
Sino Biological Recombinant Human IL-2R gamma/CD132 Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10555H49HB100
Encompass_Preferred
A DNA sequence encoding the human IL2RG (NP_000197.1) (Met1-Ala262) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4479