Learn More
Sino Biological Recombinant Anti-IL-3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11858R607100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL3 (rh IL3; Catalog#11858-H08B; NP_000579.2; Met1-Phe152).
Catalog No. 50-240-8946