Learn More
Sino Biological Anti-IL-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11858T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL3/IL-3/Interleukin-3 (rh IL3/IL-3/Interleukin-3; Catalog#11858-H08B; NP_000579.2; Met1-Phe152). IL3/IL-3/Interleukin-3 specific IgG was purified by Human IL3/IL-3/Interleukin-3 affinity chromatography.
Catalog No. 50-241-2401