Learn More
Sino Biological Recombinant Human IL-3 Protein, GMP grade, HPLC-verified
Supplier:  Sino Biological GMP11858HNAE1
Encompass_Preferred
A DNA sequence encoding the human IL3 (NP_000579.2) (Ala20-Phe152) was expressed.
Catalog No. 50-241-0181