Learn More
Sino Biological Recombinant Human IL-31 Protein (His & AVI Tag), MALS-verified
Catalog No. 502993454
Encompass_Preferred
A DNA sequence encoding the Human IL31 (NP_001014358.1) (Ser24-Thr164) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3454 Supplier Sino Biological Supplier No. 11557H49HB100
A DNA sequence encoding the Human IL31 (NP_001014358.1) (Ser24-Thr164) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.