Learn More
Proteintech Group Inc Human IL31 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0008696T
Encompass_Preferred
Interleukin-31 (IL-31) is a newly discovered four-helix bundle cytokine and expressed by several types of cells under both normal and disease state, such as activated CD4+T cells, mast cells, monocytes, macrophages, immature and mature monocyte-derived dendritic cells and so on. IL-31 signals through a heterodimeric receptor complex consisting of the IL31 receptor alpha (IL31Rα) and the oncostatin M receptor (OSMR). IL-31 plays a predominant function in the mediation of inflammatory and lymphoma-associated itch. Increased IL-31 expression has been detected in inflamed tissues from patients with atopic dermatitis, bowel diseases, allergic asthma and rhinitis.
Catalog No. 50-197-4049