Learn More
Sino Biological Anti-IL-31 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11557RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL31 (rh IL31; Catalog#11557-H08H; NP_001014358.1; Met1-Thr164). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9460