Learn More
Sino Biological Human IL31 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11557T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL31 (rh IL31; Catalog#11557-H08H; NP_001014358.1; Met1-Thr164). IL31 specific IgG was purified by Human IL31 affinity chromatography.
Catalog No. 50-237-1606