Learn More
Sino Biological Recombinant Human IL-31 Protein (His Tag)
Supplier:  Sino Biological 11557H08H1
Encompass_Preferred
A DNA sequence encoding the human IL31 (NP_001014358.1) (Met1-Thr164) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4692