Learn More
Sino Biological Recombinant Anti-IL-33 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10368R839100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-33 (rh IL-33; Catalog#10368-HNAE; NP_254274.1; Ser112-Thr270).
Catalog No. 50-241-1405